Thermo Scientific Chemicals

Platinum(IV) bromide, 99.99% (metals basis), Pt 37.1% min, Thermo Scientific Chemicals

Catalog number: 012171.03
1 g, Each
Thermo Scientific Chemicals

Platinum(IV) bromide, 99.99% (metals basis), Pt 37.1% min, Thermo Scientific Chemicals

Catalog number: 012171.03
1 g, Each
Quantity
Catalog number: 012171.03
also known as 012171-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
68938-92-1
IUPAC Name
tetrabromoplatinumtetrakis(ylium)
Molecular Formula
Br4Pt
InChI Key
DYFRBYUUSCNRBP-UHFFFAOYSA-J
SMILES
Br[Pt+4](Br)(Br)Br
Form
Powder or crystals or crystalline powder
Assay from Suppliers CofA
Pt: ≥37.1%
Total Metal Impurities
≤0.01%

Description

Platinum(IV) bromide can be used as an analytical reagent to test for the presence of cesium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text