Ruthenium(III) nitrosylnitrate, Ru 31.3% min, Thermo Scientific Chemicals
Ruthenium(III) nitrosylnitrate, Ru 31.3% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ruthenium(III) nitrosylnitrate, Ru 31.3% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number 012175.14
also known as 012175-14
Price (USD)/ Each
1,461.65
Online exclusive
1624.00 
Save 162.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
1,461.65
Online exclusive
1624.00 
Save 162.35 (10%)
Add to cart
Ruthenium(III) nitrosylnitrate, Ru 31.3% min, Thermo Scientific Chemicals
Catalog number012175.14
Price (USD)/ Each
1,461.65
Online exclusive
1624.00 
Save 162.35 (10%)
-
Add to cart
Chemical Identifiers
CAS34513-98-9
IUPAC Nameλ²-ruthenium(2+) iminooxidanium trinitrate
Molecular FormulaN4O10Ru
InChI KeyWOSOOWIGVAKGOC-UHFFFAOYSA-N
SMILES[Ru++].N#[O+].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red/brown
FormCrystalline
Assay (special)Ruthenium, Ru: ≥31.3%
Ruthenium(III) nitrosyl nitrate used as a catalyst in various reactions namely hydrogenation, carbonylation, hydrosilylation, hydrofomylation, C-X coupling (eg Heck, Suzuki, Stille) and chiral catalysis reactions. It is also used in the production of thick-film resistors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only