Thermo Scientific Chemicals

Sodium hexachloroosmate(IV) dihydrate, Os 38.7% min, Thermo Scientific Chemicals

Catalog number: 012176.03
1 g, Each
Thermo Scientific Chemicals

Sodium hexachloroosmate(IV) dihydrate, Os 38.7% min, Thermo Scientific Chemicals

Catalog number: 012176.03
1 g, Each
Quantity
Catalog number: 012176.03
also known as 012176-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1307-81-9
Assay (unspecified)
Os 38.7% min

Description

Sodium hexachloroosmate(IV) dihydrate is used as an intermediate and catalyst in organic synthesis. It is also used to prepare dihydrogen complexes of osmium(II) containing meso-tetraphos-1 ligand.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text