Dihydrogen hexachloroosmate(IV) hydrate, Premion™, 99.95% (metals basis), Os 37% min, Thermo Scientific Chemicals
Dihydrogen hexachloroosmate(IV) hydrate, Premion™, 99.95% (metals basis), Os 37% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dihydrogen hexachloroosmate(IV) hydrate, Premion™, 99.95% (metals basis), Os 37% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
0.5 g
Catalog number 012178.03
Price (USD)/ Each
144.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
144.00
Add to cart
Dihydrogen hexachloroosmate(IV) hydrate, Premion™, 99.95% (metals basis), Os 37% min, Thermo Scientific Chemicals
Catalog number012178.03
Price (USD)/ Each
144.00
-
Add to cart
Chemical Identifiers
CAS27057-71-2
IUPAC Namehexachloroosmium
Molecular FormulaH2Cl6Os
InChI KeyGCRLVKBHFZOVLQ-UHFFFAOYSA-H
SMILESCl[Os](Cl)(Cl)(Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.05% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only