Thermo Scientific Chemicals

Iridium(IV) chloride, 99.95% (metals basis), Ir 56.5 min, Thermo Scientific Chemicals

Catalog number: 012184.06
5 g, Each
Thermo Scientific Chemicals

Iridium(IV) chloride, 99.95% (metals basis), Ir 56.5 min, Thermo Scientific Chemicals

Catalog number: 012184.06
5 g, Each
Quantity
Catalog number: 012184.06
also known as 012184-06
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10025-97-5
IUPAC Name
iridium(4+) tetrachloride
Molecular Formula
Cl4Ir
InChI Key
CALMYRPSSNRCFD-UHFFFAOYSA-J
SMILES
[Cl-].[Cl-].[Cl-].[Cl-].[Ir+4]
Form
Glassy amorphous pieces/powder
Assay (special)
Ir: ≥56.5%
Total Metal Impurities
≤0.05%
Appearance (Color)
Black

Description

Iridium(IV) chloride is used in chemical analysis as testing for nitric acid in the presence of nitrous acid, microscopy and in plating solutions. It acts as a catalyst precursor for the stereoselective reduction of cyclohexanones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text