Thermo Scientific Chemicals

Molybdenum carbide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 012192.22
100 g, Each
Thermo Scientific Chemicals

Molybdenum carbide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 012192.22
100 g, Each
Quantity
Catalog number: 012192.22
also known as 012192-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12069-89-5
IUPAC Name
(molybdeniomethylidene)molybdenum
Molecular Formula
CMo2
InChI Key
QIJNJJZPYXGIQM-UHFFFAOYSA-N
SMILES
[Mo]=C=[Mo]
Form
Powder
Total Metal Impurities
≤0.5%
Appearance (Color)
Black or grey to dark grey
Assay (unspecified)
5.5-5.9% C

Description

Molybdenum carbide is used in the production of solid molybdenum carbide grades of hard metal in the manufacture of mixed crystal. It used as coating material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text