Thermo Scientific Chemicals

Barium hydroxide, anhydrous, 94-98%, Thermo Scientific Chemicals

Catalog number: 012195.30
250 g, Each
Thermo Scientific Chemicals

Barium hydroxide, anhydrous, 94-98%, Thermo Scientific Chemicals

Catalog number: 012195.30
250 g, Each
Quantity
Catalog number: 012195.30
also known as 012195-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
17194-00-2
IUPAC Name
barium(2+) dihydroxide
Molecular Formula
BaH2O2
InChI Key
RQPZNWPYLFFXCP-UHFFFAOYSA-L
SMILES
[OH-].[OH-].[Ba++]
Assay (unspecified)
94-98%

Description

Cobalt(II,III) oxide is used in enamels, pigments, porcelain colorants, semiconductors, ceramics, catalysis, grinding wheels and other fields as an important inorganic material. It is a useful oxidant in chemical industry and as catalyst for organic synthesis. It is widely used as catalyst carrier and as electrode active material. It is also used in senior goggles, other filter materials, in temperature and gas sensors, in electronic ceramics, in lithium ion battery electrode materials, magnetic materials, electrochromic devices and solar energy absorbers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text