Thermo Scientific Chemicals

Barium bromide dihydrate, 99.3%, Thermo Scientific Chemicals

Catalog number: 012197.36
500 g, Each
Thermo Scientific Chemicals

Barium bromide dihydrate, 99.3%, Thermo Scientific Chemicals

Catalog number: 012197.36
500 g, Each
Quantity
Catalog number: 012197.36
also known as 012197-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7791-28-8
IUPAC Name
barium(2+) dihydrate dibromide
Molecular Formula
BaBr2H4O2
InChI Key
HQQMRSGBIPFNSN-UHFFFAOYSA-L
SMILES
O.O.[Br-].[Br-].[Ba++]
Assay (unspecified)
99.3% min

Description

Barium bromide dihydrate is used to purify radium and make zinc bromide. It also used  precursor to chemicals used in photograph.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text