Strontium chloride, anhydrous, 95%, Thermo Scientific Chemicals
Strontium chloride, anhydrous, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Strontium chloride, anhydrous, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
Catalog number 012202.36
also known as 012202-36
Price (USD)/ Each
64.65
Online exclusive
71.30 
Save 6.65 (9%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
64.65
Online exclusive
71.30 
Save 6.65 (9%)
Add to cart
Strontium chloride, anhydrous, 95%, Thermo Scientific Chemicals
Catalog number012202.36
Price (USD)/ Each
64.65
Online exclusive
71.30 
Save 6.65 (9%)
-
Add to cart
Chemical Identifiers
CAS10476-85-4
IUPAC Namestrontium(2+) hexahydrate dichloride
Molecular FormulaCl2H12O6Sr
InChI KeyAMGRXJSJSONEEG-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Sr++]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)94.1% min
It is widely used in fireworks for the source of red color. It is also used in dental and biological research. It is the precursor material to other compounds of strontium. It is used to prepare yellow strontium chromate, a corrosion inhibitor for aluminum. It is also employed in glass-making, sea water aquaria, and metallurgy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only