Thermo Scientific Chemicals

Sodium molybdenum oxide, anhydrous, Mo 46.2%, Thermo Scientific Chemicals

Catalog number: 012209.22
100 g, Each
Thermo Scientific Chemicals

Sodium molybdenum oxide, anhydrous, Mo 46.2%, Thermo Scientific Chemicals

Catalog number: 012209.22
100 g, Each
Quantity
Catalog number: 012209.22
also known as 012209-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7631-95-0
IUPAC Name
disodium dioxomolybdenumbis(olate)
Molecular Formula
MoNa2O4
InChI Key
TVXXNOYZHKPKGW-UHFFFAOYSA-N
SMILES
[Na+].[Na+].[O-][Mo]([O-])(=O)=O
Assay from Supplier's CofA
Na₂MoO4: ≥99.2% (≥46.2% Mo Calculated)
Form
Powder

Description

It is used as an analytical reagent, corrosion inhibitor, and non-oxidizing anodic inhibitor. It reduces the nitrite requirement of fluids inhibited with nitrite-amine and improves the corrosion protection of carboxylate salt fluids. It is utilized in pigments, dyes, anti-freezing agent, cutting oils, metal finishing agent, brightening agent for zinc, additive for fertilizers and micronutrients.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text