Thermo Scientific Chemicals

Molybdenum(IV) sulfide, 98%, Thermo Scientific Chemicals

Catalog number: 012213.A1
1 kg, Each
Thermo Scientific Chemicals

Molybdenum(IV) sulfide, 98%, Thermo Scientific Chemicals

Catalog number: 012213.A1
1 kg, Each
Quantity
Catalog number: 012213.A1
also known as 012213-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1317-33-5
IUPAC Name
disulfanylidenemolybdenum
Molecular Formula
MoS2
InChI Key
CWQXQMHSOZUFJS-UHFFFAOYSA-N
SMILES
S=[Mo]=S
Assay (unspecified)
98% min

Description

It is widely used as a solid lubricant .It finds applications mainly as Intermediates, Process regulators, Processing aids. Employed as a co catalyst for desulfurization in petrochemistry. As a catalyst in the electrolysis of water.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text