Thermo Scientific Chemicals

Ammonium molybdate (di), Mo 56.5%, Thermo Scientific Chemicals

Catalog number: 012215.18
50 g, Each
Thermo Scientific Chemicals

Ammonium molybdate (di), Mo 56.5%, Thermo Scientific Chemicals

Catalog number: 012215.18
50 g, Each
Quantity
Catalog number: 012215.18
also known as 012215-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
27546-07-2
Assay (unspecified)
56.5% Mo min

Description

Ammonium molybdate (di) is used as pigments, agricultural fertilizer, color lakes and fabric fire retardant agent. It is also used as a raw material for the production of molybdenum wire, ceramic glazes and other molybdenum compounds. Further, it is used in photography, ceramics and paints. It is also used reagent for analysis as well as in the determination of phosphorus in analytical chemistry. In addition, it is used as a catalyst in the petrochemical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text