Thermo Scientific Chemicals
Platinum Rhodium Palladium gauze, 80 mesh knitted from 0.076mm (0.003in) dia wire, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number: 012216.FI
50 x 50 mm, Each
Thermo Scientific Chemicals
Platinum Rhodium Palladium gauze, 80 mesh knitted from 0.076mm (0.003in) dia wire, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number: 012216.FI
50 x 50 mm, Each
Quantity
Catalog number: 012216.FI
also known as 012216-FI
Price (USD)
Have Questions?
Total Metal Impurities
0.1% max.
Description
Platinum Rhodium Palladium gauze, 80 mesh woven from 0.076mm (0.003in) dia wire, 99.9% (metals basis) is used as a catalyst in the preparation of nitric acid from ammonia. It is also used in chemical research.