Thermo Scientific Chemicals

Platinum Rhodium gauze, 80 mesh woven from 0.076mm (0.003in) dia wire, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 012217.GH
100 x 100 mm, Each
Thermo Scientific Chemicals

Platinum Rhodium gauze, 80 mesh woven from 0.076mm (0.003in) dia wire, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 012217.GH
100 x 100 mm, Each
Quantity
Catalog number: 012217.GH
also known as 012217-GH
Price (USD)
Have Questions?
Total Metal Impurities
≤0.1%
Form
Gauze
Assay from Supplier's CofA
Pt: 89.5-90.5%
Assay from Supplier's CofA
Rh: 9.5-10.5%

Description

Platinum-rhodium alloy gauze serves as a catalyst in the oxidation of the ammonia from the Haber Process by atmospheric oxygen to produce nitric oxide and subsequently nitric acid, which is used for producing nitrogen fertilizers, ammonium nitrate, adipic acid, and toluene diisocyanate (precursor for polyurethane). It has high degree of chemical inertness and stability at very high temperatures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text