Thermo Scientific Chemicals

Lead(II) oxide, 99.9%, Thermo Scientific Chemicals

Catalog number: 012220.A3
2 kg, Each
Thermo Scientific Chemicals

Lead(II) oxide, 99.9%, Thermo Scientific Chemicals

Catalog number: 012220.A3
2 kg, Each
Quantity
Catalog number: 012220.A3
also known as 012220-A3
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1317-36-8
IUPAC Name
plumbanone
Molecular Formula
OPb
InChI Key
YEXPOXQUZXUXJW-UHFFFAOYSA-N
SMILES
O=[Pb]
Assay from Suppliers CofA
≥99.90% (unspecified)
Appearance (Color)
Pale yellow to yellow or pale orange
Form
Powder or crystalline powder or crystals

Description

It is employed mostly in lead-based industrial glass and industrial ceramics, including computer components. Lead monoxide is used as an intermediate/precursor in the manufacture of several products, for example water proof cements, lubricants, lubricating oils, inorganic pigments, lead soaps, petroleum refining, rubber, cathode ray tube glass, and polyvinyl chloride (PVC). It is useful for lead acid batteries as cathode and anode. Lead monoxide scores significant applications in oil, gas and chemical manufactures. It is an efficient catalyst for condensation reactions in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text