Thermo Scientific Chemicals

Nickel(II) acetate hydrate, 99+%, Thermo Scientific Chemicals

Catalog number: 012223.30
250 g, Each
Thermo Scientific Chemicals

Nickel(II) acetate hydrate, 99+%, Thermo Scientific Chemicals

Catalog number: 012223.30
250 g, Each
Quantity
Catalog number: 012223.30
also known as 012223-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6018-89-9
IUPAC Name
nickel(2+) diacetate tetrahydrate
Molecular Formula
C4H14NiO8
InChI Key
OINIXPNQKAZCRL-UHFFFAOYSA-L
SMILES
O.O.O.O.[Ni++].CC([O-])=O.CC([O-])=O
Appearance (Color)
Blue-green
Form
Powder or crystals or crystalline powder
Assay (special)
Nickel: ≥23.0%
Total Metal Impurities
≤1.0%

Description

Used in synthesis of polynuclear-nickel polyoxotungstate cluster compounds that are ideal for the design of molecular magnets .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text