Thermo Scientific Chemicals

Magnesium acetate tetrahydrate, 98% (metals basis), Thermo Scientific Chemicals

Catalog number: 012225.30
250 g, Each
Thermo Scientific Chemicals

Magnesium acetate tetrahydrate, 98% (metals basis), Thermo Scientific Chemicals

Catalog number: 012225.30
250 g, Each
Quantity
Catalog number: 012225.30
also known as 012225-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16674-78-5
IUPAC Name
magnesium(2+) diacetate tetrahydrate
Molecular Formula
C4H14MgO8
InChI Key
XKPKPGCRSHFTKM-UHFFFAOYSA-L
SMILES
O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O
Appearance (Color)
White
Total Metal Impurities
≤2.0%
Form
Crystals or crystalline powder

Description

Magnesium acetate tetrahydrate is commonly used as a source of magnesium in biological reactions It is an essential compound for a number of biochemical processes. Involved in glycolysis and the Krebs cycle, formation of cyclic-AMP and ATP.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text