Cobalt(II) acetate tetrahydrate, Co 24%, Thermo Scientific Chemicals
Cobalt(II) acetate tetrahydrate, Co 24%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cobalt(II) acetate tetrahydrate, Co 24%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Catalog number 012227.A1
also known as 012227-A1
Price (USD)/ Each
126.65
Online exclusive
141.00 
Save 14.35 (10%)
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
126.65
Online exclusive
141.00 
Save 14.35 (10%)
Add to cart
Cobalt(II) acetate tetrahydrate, Co 24%, Thermo Scientific Chemicals
Catalog number012227.A1
Price (USD)/ Each
126.65
Online exclusive
141.00 
Save 14.35 (10%)
-
Add to cart
Chemical Identifiers
CAS6147-53-1
IUPAC Nameλ²-cobalt(2+) diacetate tetrahydrate
Molecular FormulaC4H14CoO8
InChI KeyZBYYWKJVSFHYJL-UHFFFAOYSA-L
SMILESO.O.O.O.[Co++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red
Assay from Suppliers CofACobalt, Co: ≥23.5%
FormCrystals or powder or crystalline powder
It is used as an industrial catalyst that allow paints and varnishes to harden. Also as a precursor to various oil drying agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only