Thermo Scientific Chemicals

Cadmium nitrate tetrahydrate, 98.5% min, Thermo Scientific Chemicals

Catalog number: 012228.36
500 g, Each
Thermo Scientific Chemicals

Cadmium nitrate tetrahydrate, 98.5% min, Thermo Scientific Chemicals

Catalog number: 012228.36
500 g, Each
Quantity
Catalog number: 012228.36
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10022-68-1
IUPAC Name
cadmium(2+) bis(nitric acid)
Molecular Formula
CdH2N2O6
InChI Key
IZIPRBSCGMISKX-UHFFFAOYSA-N
SMILES
[Cd++].O[N+]([O-])=O.O[N+]([O-])=O
Assay (unspecified)
98.5% min

Description

Cadmium nitrate is used in manufacturing of cadmium salts, photographic emulsions, laboratory reagent and coloring glass. It is used for porcelain and as a flash powder in photography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text