Platinum granules, 6.35mm (0.25in) & down, 99.95% (metals basis), Thermo Scientific Chemicals
Platinum granules, 6.35mm (0.25in) & down, 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Platinum granules, 6.35mm (0.25in) & down, 99.95% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number 012236.06
also known as 012236-06
Price (USD)/ Each
1,286.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
1,286.00
Add to cart
Platinum granules, 6.35mm (0.25in) & down, 99.95% (metals basis), Thermo Scientific Chemicals
Catalog number012236.06
Price (USD)/ Each
1,286.00
-
Add to cart
Chemical Identifiers
CAS7440-06-4
IUPAC Nameplatinum
Molecular FormulaPt
InChI KeyBASFCYQUMIYNBI-UHFFFAOYSA-N
SMILES[Pt]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver to gray
Diameter6.35mm and down
Total Metal Impurities0.05% max.
FormGranules
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only