Dysprosium pieces, distilled dendritic, 99.9% (REO), Thermo Scientific Chemicals
Dysprosium pieces, distilled dendritic, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dysprosium pieces, distilled dendritic, 99.9% (REO), Thermo Scientific Chemicals

Dysprosium pieces, 99.9%, CAS Number-7429-91-6, atom, powder, pieces, foil, 3n, disprosio, chips, 3n, ion, ingot, ingot, 3n, ingot reo, 100 g, (REO), CHEBI:33377, Silver, Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S., 231-073-9, Danger
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number 012257.22
also known as 012257-22
Price (USD)/ Each
881.65
Online exclusive
979.00 
Save 97.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
881.65
Online exclusive
979.00 
Save 97.35 (10%)
Add to cart
Dysprosium pieces, distilled dendritic, 99.9% (REO), Thermo Scientific Chemicals
Catalog number012257.22
Price (USD)/ Each
881.65
Online exclusive
979.00 
Save 97.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7429-91-6
IUPAC Namedysprosium
Molecular FormulaDy
InChI KeyKBQHZAAAGSGFKK-UHFFFAOYSA-N
SMILES[Dy]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under argon
RUO – Research Use Only