Terbium pieces, distilled dendritic, 99.9% (REO), Thermo Scientific Chemicals
Terbium pieces, distilled dendritic, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Terbium pieces, distilled dendritic, 99.9% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number 012258.06
also known as 012258-06
Price (USD)/ Each
249.65
Online exclusive
277.00 
Save 27.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
249.65
Online exclusive
277.00 
Save 27.35 (10%)
Add to cart
Terbium pieces, distilled dendritic, 99.9% (REO), Thermo Scientific Chemicals
Catalog number012258.06
Price (USD)/ Each
249.65
Online exclusive
277.00 
Save 27.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-27-9
IUPAC Nameterbium
Molecular FormulaTb
InChI KeyGZCRRIHWUXGPOV-UHFFFAOYSA-N
SMILES[Tb]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under argon
RUO – Research Use Only