Thermo Scientific Chemicals

Gallium phosphide, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 012266.18
50 g, Each
Thermo Scientific Chemicals

Gallium phosphide, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 012266.18
50 g, Each
Quantity
Catalog number: 012266.18
also known as 012266-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12063-98-8
Total Metal Impurities
0.001% max.

Description

Gallium phosphide (GaP) is used in semiconductor devices, and in the manufacture of light-emitting diodes (LEDs) doped with other elements or in combination with gallium arsenide phosphide. With appropriate dopants, p-type and n-type semiconductors can be produced. It is used in optical systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text