Thermo Scientific Chemicals

Mercury(II) acetate, ACS, 98.0% min, Thermo Scientific Chemicals

Catalog number: 012273.18
50 g, Each
Thermo Scientific Chemicals

Mercury(II) acetate, ACS, 98.0% min, Thermo Scientific Chemicals

Catalog number: 012273.18
50 g, Each
Quantity
Catalog number: 012273.18
also known as 012273-18
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1600-27-7
IUPAC Name
mercury(2+) diacetate
Molecular Formula
C4H6HgO4
InChI Key
BRMYZIKAHFEUFJ-UHFFFAOYSA-L
SMILES
[Hg++].CC([O-])=O.CC([O-])=O
Impurity content
SO₄ 0.005%
Insoluble material
0.01%
Trace Metals
(as Pb) 0.002%
Assay (unspecified)
98.0 C₄H₆HgO₄ min
Comment
Residue after reduction 0.02%

Description

Mercuric acetate is used as an oxidizing agent in organic synthesis. It is used in oxymercuration of double bonds. Mercuric acetate is used in non-aqueous titration. It is employed in the manufacture of phenyl mercury compounds which have pharmaceutical applications. It removes the acetamidomethyl protecting group from protected thiol, and converts thiocarbonate esters into dithiocarbonates. It promotes the addition of hydroxide and alkoxide across carbon-carbon double bonds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text