Thermo Scientific Chemicals

Tellurium(IV) chloride, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 012284.14
25 g, Each
Thermo Scientific Chemicals

Tellurium(IV) chloride, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 012284.14
25 g, Each
Quantity
Catalog number: 012284.14
also known as 012284-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10026-07-0
IUPAC Name
tetrachloro-λ⁴-tellane
Molecular Formula
Cl4Te
InChI Key
SWLJJEFSPJCUBD-UHFFFAOYSA-N
SMILES
Cl[Te](Cl)(Cl)Cl
Total Metal Impurities
0.1% max.

Description

Tellurium(IV) chloride is used in organic synthesis to prepare Cl-C-C-TeCl3derivatives by reacting with alkenes and aryl tellurium compounds by reacting with electron rich arenes. It is involved in the preparation of diaryl telluride from anisole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text