Thermo Scientific Chemicals

Chromium(III) oxide, 98+%, Thermo Scientific Chemicals

Catalog number: 012286.36
500 g, Each
Thermo Scientific Chemicals

Chromium(III) oxide, 98+%, Thermo Scientific Chemicals

Catalog number: 012286.36
500 g, Each
Quantity
Catalog number: 012286.36
also known as 012286-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1308-38-9
IUPAC Name
dichromium(3+) trioxidandiide
Molecular Formula
Cr2O3
InChI Key
UOUJSJZBMCDAEU-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Cr+3].[Cr+3]
Assay (unspecified)
98.1% min

Description

It is employed in metallurgical processing and has applications in chrome plating, anodizing aluminum etc… It is also used as a pigment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text