Magnesium chloride hexahydrate, 97+%, Thermo Scientific Chemicals
Magnesium chloride hexahydrate, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Magnesium chloride hexahydrate, 97+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
500 g
10 kg
Catalog number 012288.A3
also known as 012288-A3
Price (USD)/ Each
66.65
Online exclusive
73.70 
Save 7.05 (10%)
-
Add to cart
Quantity:
2 kg
Request bulk or custom format
Price (USD)/ Each
66.65
Online exclusive
73.70 
Save 7.05 (10%)
Add to cart
Magnesium chloride hexahydrate, 97+%, Thermo Scientific Chemicals
Catalog number012288.A3
Price (USD)/ Each
66.65
Online exclusive
73.70 
Save 7.05 (10%)
-
Add to cart
Chemical Identifiers
CAS7791-18-6
IUPAC Namemagnesium(2+) hexahydrate dichloride
Molecular FormulaCl2H12MgO6
InChI KeyDHRRIBDTHFBPNG-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Mg++].[Cl-].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder or crystals or crystalline powder
Assay from Suppliers CofAMagnesium Chloride (MgCl₂): ≥45.4% (≥97.0% purity calculated as hexahydrate)
Impurity contentCalcium: Report %
Impurity contentPotassium: Report %
View more
It is utilized in the polymerase chain reactions (PCR). It is used for dust control, ice control, and road stabilization. It is employed in a variety of applications such as fertilizers, mineral supplements for animals, waste water treatment, artificial seawater, textiles, paper, fireproofing agents, cements, and fire extinguishers. Pharmaceutical preparations are used in several medicines and topical or skin related applications. It acts as a coagulant in the preparation of tofu from soy milk.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only