Mercury(II) iodide, ACS, 99.0% min (Assay-dried basis), Thermo Scientific Chemicals
Mercury(II) iodide, ACS, 99.0% min (Assay-dried basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Mercury(II) iodide, ACS, 99.0% min (Assay-dried basis), Thermo Scientific Chemicals

For Use As An Analytical Reagent For the Preparation of Nessler'S Reagent.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 012289.36
also known as 012289-36
Price (USD)/ Each
398.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
469.00 
Save 70.35 (15%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
398.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
469.00 
Save 70.35 (15%)
Add to cart
Mercury(II) iodide, ACS, 99.0% min (Assay-dried basis), Thermo Scientific Chemicals
Catalog number012289.36
Price (USD)/ Each
398.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
469.00 
Save 70.35 (15%)
-
Add to cart
Chemical Identifiers
CAS7774-29-0
IUPAC Namemercury(2+) diiodide
Molecular FormulaHgI2
InChI KeyYFDLHELOZYVNJE-UHFFFAOYSA-L
SMILES[I-].[I-].[Hg++]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99.0% HgI₂ min
CommentSoluble mercury salts (as Hg) 0.05%
Impurity contentMercurous mercury (as Hg) 0.1%
Solution Testin potassium iodide solution P.T.
As an analytical reagent for the preparation of Nessler's Reagent. It is also used as an oxidizing agent for the synthesis of quinoxaline. Polycrystalline films of mercury (II) iodide, also play an important role in X-ray imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only