Thermo Scientific Chemicals

Copper(II) oxide, 97%, Thermo Scientific Chemicals

Catalog number: 012299.A9
10 kg, Each
Thermo Scientific Chemicals

Copper(II) oxide, 97%, Thermo Scientific Chemicals

Catalog number: 012299.A9
10 kg, Each
Quantity
Catalog number: 012299.A9
also known as 012299-A9
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1317-38-0
IUPAC Name
copper(2+) oxidandiide
Molecular Formula
CuO
InChI Key
KKCXRELNMOYFLS-UHFFFAOYSA-N
SMILES
[O--].[Cu++]
Assay (unspecified)
96.1% min

Description

As a pigment, in sweeting petroleum gases, in galvanic electrodes, metallurgical flux, as an optical glass polishing agent, and as a catalyst in organic reactionsCopper(II) oxide is used as an adsorbent and absorbent, pigment, antifouling agent, wood preservative, optical glass polishing agent, welding flux for bronze materials, agricultural chemical and abrasive. It is used in the preparation of intermediates, oxidizing/reducing agents, surface treating agents and process regulators. It finds use in the manufacture of rayons, phosphors, and dry battery cells. It serves as a catalyst in certain chemical reactions of industrial importance. It is also employed in chemical sensors and superconducting materials. Hazardous materials like cyanides, dioxins and halogenated hydrocarbons can be disposed through oxidation with copper oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text