Cobalt(II) chloride, anhydrous, 99.7% (metals basis), Thermo Scientific Chemicals
Cobalt(II) chloride, anhydrous, 99.7% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cobalt(II) chloride, anhydrous, 99.7% (metals basis), Thermo Scientific Chemicals

Sensitivity: Hygroscopic; 3.367. Decomposes at 400°C on long heating in air
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 012303.18
also known as 012303-18
Price (USD)/ Each
71.65
Online exclusive
79.70 
Save 8.05 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
71.65
Online exclusive
79.70 
Save 8.05 (10%)
Add to cart
Cobalt(II) chloride, anhydrous, 99.7% (metals basis), Thermo Scientific Chemicals
Catalog number012303.18
Price (USD)/ Each
71.65
Online exclusive
79.70 
Save 8.05 (10%)
-
Add to cart
Chemical Identifiers
CAS7646-79-9
IUPAC Nameλ²-cobalt(2+) dichloride
Molecular FormulaCl2Co
InChI KeyGVPFVAHMJGGAJG-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Co++]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.3% max.
Cobalt(II) chloride is used in humidity indicator in weather instruments. In the anhydrous form, it finds use in electroplating of cobalt, in organic chemistry and is a precursor to cobaltocene, (bis(cyclopentadienyl)cobalt(II), which is a good reducing agent. It also serves as a Lewis acid. Cobalt chloride is an indicator for water in desiccants, owing to the reversible hydration/dehydration coupled with distinct color change. Cobalt chloride is useful for producing invisible ink as it turns blue when heated and becomes invisible once it gets cooled. Cobalt(II) chloride catalyzes cross coupling of aryl halides or vinyl halides with aryl grignard reagents in excellent yields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only