Thermo Scientific Chemicals

Barium chloride dihydrate, ACS, Thermo Scientific Chemicals

Catalog number: 012310.36
500 g, Each
Thermo Scientific Chemicals

Barium chloride dihydrate, ACS, Thermo Scientific Chemicals

Catalog number: 012310.36
500 g, Each
Quantity
Catalog number: 012310.36
also known as 012310-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10326-27-9
IUPAC Name
barium(2+) dihydrate dichloride
Molecular Formula
BaCl2H4O2
InChI Key
PWHCIQQGOQTFAE-UHFFFAOYSA-L
SMILES
O.O.[Cl-].[Cl-].[Ba++]
Appearance (Color)
White
Form
Crystals or crystalline powder
Assay from Supplier's CofA
≥99.0% BaCl₂.2H₂O
Heavy Metals
(ICP-OES): ≤5ppm
Impurity content
Ca: ≤0.05%

Description

Barium chloride is usually employed as a chemical reagent to precipitate soluble sulfates and also in the purification of the seawater solution. It is practiced in several applications like heat-treating baths, waste water treatment, the production of PVC stabilizers, oil lubricants, fireworks materials, hardening of steel, in the manufacture of pigments and in the manufacture of other barium salts. It is applied to prepare composites to absorb ammonia and to increase the crystallinty of polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text