Thermo Scientific Chemicals

Calcium chloride dihydrate, 99% min, Thermo Scientific Chemicals

Catalog number: 012312.A3
2 kg, Each
Thermo Scientific Chemicals

Calcium chloride dihydrate, 99% min, Thermo Scientific Chemicals

Catalog number: 012312.A3
2 kg, Each
Quantity
Catalog number: 012312.A3
also known as 012312-A3
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10035-04-8
IUPAC Name
calcium dihydrate dichloride
Molecular Formula
CaCl2H4O2
InChI Key
LLSDKQJKOVVTOJ-UHFFFAOYSA-L
SMILES
O.O.[Cl-].[Cl-].[Ca++]
Appearance (Color)
White
Assay (special)
99.0-107.0%
Form
Granular

Description

Calcium chloride dihydrate is widely used as an ingredient in ceramic slipware, cosmetic and food items. It acts as a pH buffer in swimming pool water. It provides inhibition of swelling clays in the water phase of invert emulsion drilling fluids. It is employed in the oil industry to increase the density of solids-free brines. It is used as a flux in the Davy process for the manufacture of sodium through the electrolysis of molten sodium chloride. It finds an important application as an additive in plastics, fire extinguishers, blast furnaces and in wastewater treatment as a drainage aid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text