Thermo Scientific Chemicals

Zinc nitrate hexahydrate, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 012313.36
500 g, Each
Thermo Scientific Chemicals

Zinc nitrate hexahydrate, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 012313.36
500 g, Each
Quantity
Catalog number: 012313.36
also known as 012313-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10196-18-6
IUPAC Name
zinc(2+) hexahydrate dinitrate
Molecular Formula
H12N2O12Zn
InChI Key
JGPSMWXKRPZZRG-UHFFFAOYSA-N
SMILES
O.O.O.O.O.O.[Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Total Metal Impurities
1% max.

Description

Zinc nitrate hexahydrate is used in the synthesis of coordination polymers. It is utilized in the preparation of zinc oxide based nanowires. It is also used as a mordant in dyeing. It is involved in the preparation of a new microporous, organically templated zinc phosphate, which finds application as catalysts and other applications involving microporosity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text