Magnesium chloride, anhydrous, 99%, Thermo Scientific Chemicals
Magnesium chloride, anhydrous, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Magnesium chloride, anhydrous, 99%, Thermo Scientific Chemicals

For Use As a Catalyst, Flocculating Agent, Various Magnesium Cements
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
1 kg
25 kg
Catalog number 012315.A7
also known as 012315-A7
Price (USD)/ Each
129.65
Online exclusive
144.00 
Save 14.35 (10%)
-
Add to cart
Quantity:
5 kg
Request bulk or custom format
Price (USD)/ Each
129.65
Online exclusive
144.00 
Save 14.35 (10%)
Add to cart
Magnesium chloride, anhydrous, 99%, Thermo Scientific Chemicals
Catalog number012315.A7
Price (USD)/ Each
129.65
Online exclusive
144.00 
Save 14.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7786-30-3
IUPAC Namemagnesium(2+) dichloride
Molecular FormulaCl2Mg
InChI KeyTWRXJAOTZQYOKJ-UHFFFAOYSA-L
SMILES[Mg++].[Cl-].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder or crystals or crystalline powder
Assay from Supplier's CofAMagnesium Chloride (MgCl₂): ≥98.5%
CommentCalcium (Ca): ≤20 ppm
CommentMagnesium Oxide (MgO): ≤2000 ppm
View more

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Hygroscopic; May contain trace amount of Zr as black insoluble particles
RUO – Research Use Only