Spectroflux 106, Lithium tetraborate & Lanthanum oxide, 85:15 w/w%
Spectroflux 106, Lithium tetraborate & Lanthanum oxide, 85:15 w/w%
Thermo Scientific Chemicals
Spectroflux 106, Lithium tetraborate & Lanthanum oxide, 85:15 w/w%
Intermediate activity flux
Have Questions?
Catalog NumberQuantity
012321.A1
also known as 012321-A1
1 kg
Catalog number 012321.A1
also known as 012321-A1
Price (USD)
712.00
Each
Add to cart
Quantity:
1 kg
Price (USD)
712.00
Each
Add to cart
Non-oxidizing, intermediate activity flux with heavy absorber. Samples must be fully oxidized. Often used in testing of steel
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Non-oxidizing, intermediate activity flux with heavy absorber. Samples must be fully oxidized. Often used in testing of steel
Specifications
Melting Point900°C
Physical FormSolid
Quantity1 kg
Unit SizeEach
Share catalog number, name or link