Thermo Scientific Chemicals

Hydrogen tetrachloroaurate(III) hydrate, 99.9% (metals basis), Au 49% min, Thermo Scientific Chemicals

Catalog number: 012325.03
1 g, Each
Thermo Scientific Chemicals

Hydrogen tetrachloroaurate(III) hydrate, 99.9% (metals basis), Au 49% min, Thermo Scientific Chemicals

Catalog number: 012325.03
1 g, Each
Quantity
Catalog number: 012325.03
also known as 012325-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
27988-77-8
IUPAC Name
hydrogen tetrachlorogoldtris(ylium)
Molecular Formula
AuCl4H
InChI Key
RFWCEGCWRSNAGT-UHFFFAOYSA-K
SMILES
[H+].Cl[Au+3](Cl)(Cl)Cl
Appearance (Color)
Yellow to orange
Form
Crystalline powder or granular powder
Assay (special)
Au 49.0-50.0%
Total Metal Impurities
≤0.1%

Description

Hydrogen tetrachloroaurate(III) hydrate is used for microanalysis of rubidium, cesium and determination of alkaloids. Usually used as a harbinger for the preparation of gold (Au) nanoparticles. It is used in control rods in nuclear reactors and in the manufacture of light electric light filaments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text