Thermo Scientific Chemicals

Tungsten(VI) chloride, 99%, Thermo Scientific Chemicals

Catalog number: 012337.06
5 g, Each
Thermo Scientific Chemicals

Tungsten(VI) chloride, 99%, Thermo Scientific Chemicals

Catalog number: 012337.06
5 g, Each
Quantity
Catalog number: 012337.06
also known as 012337-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13283-01-7
IUPAC Name
tungsten hexachloride
Molecular Formula
Cl6H6W
InChI Key
XHYHOBJDYHRWHO-UHFFFAOYSA-N
SMILES
Cl.Cl.Cl.Cl.Cl.Cl.[W]
Assay (unspecified)
98.6% min

Description

It is a precursor for many tungsten compounds. It is used in tungsten plating, purification of tungsten, and olefin polymerization. It is also used as petroleum catalyst, and in intelligent glass. It can be used for deoxygenation of epoxides and as a reagent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text