Thermo Scientific Chemicals

Cadmium sulfate octahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals

Catalog number: 012340.30
250 g, Each
Thermo Scientific Chemicals

Cadmium sulfate octahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals

Catalog number: 012340.30
250 g, Each
Quantity
Catalog number: 012340.30
also known as 012340-30
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7790-84-3
IUPAC Name
cadmium(2+) hydrate sulfate
Molecular Formula
CdH2O5S
InChI Key
VRFRVKRTIMIRNG-UHFFFAOYSA-L
SMILES
O.[Cd++].[O-]S([O-])(=O)=O
Impurity content
Cl 0.001%
Impurity content
Pb 0.003%
Impurity content
Cu 0.002%
Assay (unspecified)
98.0-102.0% 3CdSO₄.8H₂O
Impurity content
Fe 0.001%

Description

Cadmium sulfate mainly used in electroplating of cadmium, which is used in electronic circuits. It is used as a precursor for cadmium based pigments such as cadmium sulfide and pharmaceutical intermediates. It is used in the standard weston cell.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text