Thermo Scientific Chemicals

Barium titanium oxide, 99%, Thermo Scientific Chemicals

Catalog number: 012348.36
500 g, Each
Thermo Scientific Chemicals

Barium titanium oxide, 99%, Thermo Scientific Chemicals

Catalog number: 012348.36
500 g, Each
Quantity
Catalog number: 012348.36
also known as 012348-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12047-27-7
IUPAC Name
barium(2+) oxotitaniumbis(olate)
Molecular Formula
BaO3Ti
InChI Key
WNKMTAQXMLAYHX-UHFFFAOYSA-N
SMILES
[Ba++].[O-][Ti]([O-])=O
Particle Size
+325 Mesh Screen: ≤0.10%
Assay from Supplier's CofA
(Ba+Sr)O: ≥65.0%
Composition
(Ba+Sr)/Ti Ratio: 0.988-1.002
Form
Powder
Assay from Supplier's CofA
TiO₂: ≥34.0%

Description

Barium titanium oxide is a dielectric ceramic used for capacitors. It is a piezoelectric material for microphones and other transducers. Finds applications in nonlinear optics and photorefractive applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text