Lead(II) chloride, Reagent Grade, 99%, Thermo Scientific Chemicals
Lead(II) chloride, Reagent Grade, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lead(II) chloride, Reagent Grade, 99%, Thermo Scientific Chemicals

Analytical Reagent, Preparation of Lead Salts, As Solder and Flux
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
500 g
2 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 012349.18
also known as 012349-18
Price (USD)/ Each
22.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
26.20 
Save 3.55 (14%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
22.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
26.20 
Save 3.55 (14%)
Add to cart
Lead(II) chloride, Reagent Grade, 99%, Thermo Scientific Chemicals
Catalog number012349.18
Price (USD)/ Each
22.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
26.20 
Save 3.55 (14%)
-
Add to cart
Chemical Identifiers
CAS7758-95-4
IUPAC Nameλ²-lead(2+) dichloride
Molecular FormulaCl2Pb
InChI KeyHWSZZLVAJGOAAY-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Pb++]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.6% min
It is employed in the manufacture of infrared transmitting glass, and ornamental glass called arena glass. It also finds application as analytical reagent, in preparation of lead salts, as solder and flux, for provision of lead titanate, and used in the purification process of ores.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only