Thermo Scientific Chemicals

Lead(II) chloride, Reagent Grade, 99%, Thermo Scientific Chemicals

Catalog number: 012349.A3
2 kg, Each
Thermo Scientific Chemicals

Lead(II) chloride, Reagent Grade, 99%, Thermo Scientific Chemicals

Catalog number: 012349.A3
2 kg, Each
Quantity
Catalog number: 012349.A3
also known as 012349-A3
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7758-95-4
IUPAC Name
λ²-lead(2+) dichloride
Molecular Formula
Cl2Pb
InChI Key
HWSZZLVAJGOAAY-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].[Pb++]
Assay (unspecified)
98.6% min

Description

It is employed in the manufacture of infrared transmitting glass, and ornamental glass called arena glass. It also finds application as analytical reagent, in preparation of lead salts, as solder and flux, for provision of lead titanate, and used in the purification process of ores.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text