Ruthenium black, 99.9% (metals basis), Thermo Scientific Chemicals
Ruthenium black, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ruthenium black, 99.9% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number 012354.04
also known as 012354-04
Price (USD)/ Each
280.65
Online exclusive
312.00 
Save 31.35 (10%)
-
Add to cart
Quantity:
2 g
Request bulk or custom format
Price (USD)/ Each
280.65
Online exclusive
312.00 
Save 31.35 (10%)
Add to cart
Ruthenium black, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number012354.04
Price (USD)/ Each
280.65
Online exclusive
312.00 
Save 31.35 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialRuthenium black
CAS7440-18-8
Health Hazard 1H228-H500
Health Hazard 2GHS H Statement
H228
Flammable solid.
Health Hazard 3P210-P240-P241-P280-P370+P378q
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only