Thermo Scientific Chemicals

Nickel(II) oxide, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 012359.30
250 g, Each
Thermo Scientific Chemicals

Nickel(II) oxide, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 012359.30
250 g, Each
Quantity
Catalog number: 012359.30
also known as 012359-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1313-99-1
IUPAC Name
oxonickel
Molecular Formula
NiO
InChI Key
GNRSAWUEBMWBQH-UHFFFAOYSA-N
SMILES
O=[Ni]
Total Metal Impurities
≤ 1%
Particle Size
-325 Mesh
Form
Powder
Appearance (Color)
Green

Description

It is used in nickel-iron battery (Edison Battery) and fuel cells. It is a precursor for many nickel salts and nickel alloys including nickel-steel allys, for example invar and elinvar. It is used in electrochromic devices, frits, ferrites, and porcelain glazes. It also serves as a hydrogenation catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text