Thermo Scientific Chemicals

Aluminum nitrate nonahydrate, 98%, Thermo Scientific Chemicals

Catalog number: 012360.30
250 g, Each
Thermo Scientific Chemicals

Aluminum nitrate nonahydrate, 98%, Thermo Scientific Chemicals

Catalog number: 012360.30
250 g, Each
Quantity
Catalog number: 012360.30
also known as 012360-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7784-27-2
IUPAC Name
aluminium(3+) nonahydrate trinitrate
Molecular Formula
AlH18N3O18
InChI Key
SWCIQHXIXUMHKA-UHFFFAOYSA-N
SMILES
O.O.O.O.O.O.O.O.O.[Al+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Assay (unspecified)
97.5% min

Description

It finds use in tanning leather, antiperspirant, corrosion protection, extraction of uranium, nitrating agent. It is an effective dopant for producing highly conductive thin film by utilizing the sol gel method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text