Ammonium chloride, 99.5% min, Thermo Scientific Chemicals
Ammonium chloride, 99.5% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium chloride, 99.5% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
Catalog number 012361.36
also known as 012361-36
Price (USD)/ Each
43.65
Online exclusive
48.80 
Save 5.15 (11%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
43.65
Online exclusive
48.80 
Save 5.15 (11%)
Add to cart
Ammonium chloride, 99.5% min, Thermo Scientific Chemicals
Catalog number012361.36
Price (USD)/ Each
43.65
Online exclusive
48.80 
Save 5.15 (11%)
-
Add to cart
Chemical Identifiers
CAS12125-02-9
IUPAC Nameamine hydrochloride
Molecular FormulaClH4N
InChI KeyNLXLAEXVIDQMFP-UHFFFAOYSA-N
SMILESN.Cl
View more
SpecificationsSpecification SheetSpecification Sheet
FormGranular
Assay (unspecified)99.5% min
Appearance (Color)White to pale yellow
It act as a buffer, as a flux in galvanizing steel sheet. It is used in batteries, to clean soldering irons, analytical lab reagent, in washing powders, electroplating, lustering cotton.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only