Thermo Scientific Chemicals

Aluminum sulfate hydrate, 97+%, Thermo Scientific Chemicals

Catalog number: 012362.A7
5 kg, Each
Thermo Scientific Chemicals

Aluminum sulfate hydrate, 97+%, Thermo Scientific Chemicals

Catalog number: 012362.A7
5 kg, Each
Quantity
Catalog number: 012362.A7
also known as 012362-A7
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
17927-65-0
IUPAC Name
dialuminium(3+) trisulfate
Molecular Formula
Al2O12S3
InChI Key
DIZPMCHEQGEION-UHFFFAOYSA-H
SMILES
[Al+3].[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Appearance (Color)
White to pale cream
Form
Crystals or crystalline powder
Assay from Supplier's CofA
≥96.0%

Description

Aluminum sulfate hydrate is a chemical compound commonly used as a flocculating agent in the purification of drinking water and waste water treatment plants, and also in paper manufacturing. Used as coagulant in decolorization and COD removal of secondary yeast wastewater effluents. It is also plays a significant role in arsenic removal by coagulation process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text