Thermo Scientific Chemicals

Ammonium nitrate, ACS, 95% min, Thermo Scientific Chemicals

Catalog number: 012363.A3
2 kg, Each
Thermo Scientific Chemicals

Ammonium nitrate, ACS, 95% min, Thermo Scientific Chemicals

Catalog number: 012363.A3
2 kg, Each
Quantity
Catalog number: 012363.A3
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6484-52-2
IUPAC Name
ammonium nitrate
Molecular Formula
H4N2O3
InChI Key
DVARTQFDIMZBAA-UHFFFAOYSA-O
SMILES
[NH4+].[O-][N+]([O-])=O
Impurity content
Fe 2ppm
Trace Metals
(as Pb) 5ppm
Assay (unspecified)
95.0% H₄N₂O₃ min
Impurity content
Cl 5ppm
pH
of a 5% solution 4.5-6.0 at 25?

Description

It is used in agriculture as a high-nitrogen fertilizer. It is one of the main components of ammonium nitrate/fuel oil (ANFO). ANFO is used in coal mining, quarrying, metal mining, hair dyes, colorings, freeaing mixtures, and civil construction. It is also used as an oxidizer in solid rocket propellants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text