Calcium nitrate tetrahydrate, ACS, 99.0-103.0%, Thermo Scientific Chemicals
Calcium nitrate tetrahydrate, ACS, 99.0-103.0%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Calcium nitrate tetrahydrate, ACS, 99.0-103.0%, Thermo Scientific Chemicals

Manufacturing of other nitrates, fertilizers, matches
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 012364.36
also known as 012364-36
Price (USD)/ Each
47.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
56.30 
Save 8.65 (15%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
47.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
56.30 
Save 8.65 (15%)
Add to cart
Calcium nitrate tetrahydrate, ACS, 99.0-103.0%, Thermo Scientific Chemicals
Catalog number012364.36
Price (USD)/ Each
47.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
56.30 
Save 8.65 (15%)
-
Add to cart
Chemical Identifiers
CAS13477-34-4
IUPAC Namecalcium bis(nitric acid) tetrahydrate
Molecular FormulaCaH10N2O10
InChI KeyDKCJOKYHYIAZQH-UHFFFAOYSA-N
SMILESO.O.O.O.[Ca++].O[N+]([O-])=O.O[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99.0-103.0% Ca(NO₃)₂?4H₂O
Heavy Metals(ICP-OES) 5ppm
Impurity contentK 0.005%
pHof a 5% solution 5.0-7.0 at 25?
Impurity contentBa 0.005%
View more
Calcium nitrate tetrahydrate is used as nitrogenous fertilizer and corrosive inhibitor in diesel fuels. It is utilized in plant cell for culture experiments. It is also used in cooling baths and waste water handling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only