Thermo Scientific Chemicals

Aluminum hydroxide, 76.5% min, Thermo Scientific Chemicals

Catalog number: 012366.A3
2 kg, Each
Thermo Scientific Chemicals

Aluminum hydroxide, 76.5% min, Thermo Scientific Chemicals

Catalog number: 012366.A3
2 kg, Each
Quantity
Catalog number: 012366.A3
also known as 012366-A3
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
21645-51-2
IUPAC Name
aluminium(3+) trihydroxide
Molecular Formula
AlH3O3
InChI Key
WNROFYMDJYEPJX-UHFFFAOYSA-K
SMILES
[OH-].[OH-].[OH-].[Al+3]
Appearance (Color)
White
Chloride content
≤0.85%
pH
≤10.0
Form
Powder, amorphous
Assay (special)
Acid Neutralizing Capacity: ≥25.0meq/g

Description

As absorbent, in chromatography, manufacturing of glass, paper, inks, ceramics, lubricants, cosmeticsAluminum hydroxide is used as a desiccant powder, filler in paper, plastics, rubber and cosmetics. It is used as a smoke suppressant and mordant dye. It is also used in drugs as antacid and antihyperphosphatemic. It is also used as a Claus catalyst support for waterproof fabrics. It is an important starting material for the preparation of other aluminum compounds, calcined alumina, aluminum sulfate, polyaluminum chloride, zeolites, sodium aluminate, activated alumina and aluminum nitrate. In addition, it is used as a fire retardant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text