Cadmium chloride hemipentahydrate, ACS, 79.5-81.0%, Thermo Scientific Chemicals
Cadmium chloride hemipentahydrate, ACS, 79.5-81.0%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cadmium chloride hemipentahydrate, ACS, 79.5-81.0%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 012373.30
also known as 012373-30
Price (USD)/ Each
102.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
120.00 
Save 17.35 (14%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
102.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
120.00 
Save 17.35 (14%)
Add to cart
Cadmium chloride hemipentahydrate, ACS, 79.5-81.0%, Thermo Scientific Chemicals
Catalog number012373.30
Price (USD)/ Each
102.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
120.00 
Save 17.35 (14%)
-
Add to cart
Chemical Identifiers
CAS7790-78-5
IUPAC Namecadmium(2+) dichloride
Molecular FormulaCdCl2
InChI KeyYKYOUMDCQGMQQO-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Cd++]
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentCa 0.005%
Impurity contentFe 5ppm
Impurity contentNa 0.05%
Impurity contentPb 0.005%
Impurity contentNitrate and nitrite (as NO₃)0.003%
View more
Cadmium chloride is also used for photocopying, dyeing and electroplating. Useful in pyrotechnic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only